missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Troglitazone

Product Code. 17190282
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17190282 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17190282 Supplier STEMCELL Technologies Supplier No. 73892
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

NF-kB pathway inhibitor; Activates PPARγ

  • Troglitazone is a potent and selective activator of peroxisome proliferator-activated receptor-γ (PPARγ) with EC50 = 0.55 and 0.78 μM for human and mouse, respectively
  • Troglitazone does not inhibit PPARα or PPARδ at up to 10 μM (Willson et al.) Troglitazone causes cell cycle arrest at phase G1, inducing caspase-dependent apoptosis in both HeLa and hepatocellular carcinoma cell lines (Yoshizawa et al.; Chang et al.)

Specifications

CAS 97322-87-7
Synonym Resulin; Rezulin
Quantity 5 mg
Molecular Formula C24H27NSO5
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.