missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Trolox

Product Code. 18044652
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18044652 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18044652 Supplier R&D Systems™ Supplier No. 6002/100
  • ONLINE EXCLUSIVE PRICE
    72.00 €
    68.25 € / 100mg
    Save 3.75 €
    5% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Antifade reagent; antioxidant vitamin E derivative; cell permeable and water soluble

Trolox is an antioxidant vitamin E derivative; suppresses osteoclast formation and inhibits IL-1-induced osteoclast bone loss. Also inhibits ferroptosis. Commonly used antifade reagent in live cell imaging; suppresses blinking and photobleaching. Cell-permeable and water soluble.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dihydro-6-hydroxy-2,5,7,8-tetramethyl-2H-1-benzopyran-2-carboxylic acid
CAS 53188-07-1
Quantity 100 mg
Product Line Tocris
Target Additional Antioxidant Compounds
Molecular Formula C14H18O4
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.