missing translation for 'onlineSavingsMsg'
Learn More

Trolox, MedChemExpress

Product Code. 16401536
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16401536 500 mg 500mg
16480759 1 g 1g
16446097 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16401536 Supplier Medchem Express Supplier No. HY101445

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trolox is an analogue of vitamin E with a powerful antioxidant effect. Trolox is also a powerful inhibitor of membrane damage.

Trolox is an analogue of vitamin E with a powerful antioxidant effect. Trolox is also a powerful inhibitor of membrane damage.

Specifications

Chemical Name or Material Trolox
CAS 53188-07-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H18O4
Quantity 500 mg
Solubility Information DMSO : ≥ 100 mg/mL (399.54 mM)
SMILES O=C(C1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1)O
Molecular Weight (g/mol) 250.29
Formula Weight 250.29
Percent Purity 98.19%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.