missing translation for 'onlineSavingsMsg'
Learn More

Trolox, MedChemExpress

Product Code. p-200101304
Encompass_Preferred
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
This item is not returnable. View return policy

Product Code. 16480759

Brand: Medchem Express HY101445

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trolox is an analogue of vitamin E with a powerful antioxidant effect. Trolox is also a powerful inhibitor of membrane damage.

Trolox is an analogue of vitamin E with a powerful antioxidant effect. Trolox is also a powerful inhibitor of membrane damage.

Specifications

Chemical Name or Material Trolox
CAS 53188-07-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H18O4
Quantity 1 g
Solubility Information DMSO : ≥ 100 mg/mL (399.54 mM)
SMILES O=C(C1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1)O
Molecular Weight (g/mol) 250.29
Formula Weight 250.29
Percent Purity 98.19%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.