missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tropaeolin O

Product Code. 11378977
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378977 2 g 2g
11388977 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378977 Supplier Thermo Scientific Chemicals Supplier No. 038708.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 547-57-9 | C12H10N2NaO5S | 317.271 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-57-9
Molecular Formula C12H10N2NaO5S
Molecular Weight (g/mol) 317.271
MDL Number MFCD00007499
InChI Key YOGUPPQYRWYKGT-IRIIKGHASA-N
Synonym Acid Orange 6; C.I. 14270
PubChem CID 87071197
IUPAC Name 4-[(2Z)-2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzenesulfonic acid;sodium
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C=C2O)S(=O)(=O)O.[Na]

Specifications

Merck Index 14,9775
Solubility Information Soluble in water and alcohol
Formula Weight 316.27
Color Brown to Orange
Quantity 2 g
Physical Form Powder
Chemical Name or Material Tropaeolin O

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.