missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tropaeolin O, pure

Product Code. 10236250 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10236250 25 g Glass bottle 25g
10688434 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10236250 Supplier Thermo Scientific Chemicals Supplier No. 190180250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 547-57-9 | C12H10N2NaO5S | 317.271 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-57-9
Molecular Formula C12H10N2NaO5S
Molecular Weight (g/mol) 317.271
MDL Number MFCD00007499
InChI Key YOGUPPQYRWYKGT-IRIIKGHASA-N
Synonym Acid orange 6, C.I. 14270, 4-(2, 4-Dihydroxyphenylazo)benzenesulfonic acid, sodium salt
PubChem CID 87071197
IUPAC Name 4-[(2Z)-2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzenesulfonic acid;sodium
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C=C2O)S(=O)(=O)O.[Na]

Specifications

Beilstein 16, 275
Merck Index 15, 9954
Solubility Information Solubility in water: Soluble. Other solubilities: Soluble in alcohol
Infrared Spectrum Authentic
Formula Weight 316.26
Grade Extra Pure
Packaging Glass bottle
Visual Transition Interval from pH 11.1 (yellow) to pH 12.7 (orange)
Color Brown to Rust
Quantity 25 g
Physical Form Powder
Chemical Name or Material Tropaeolin O
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.