missing translation for 'onlineSavingsMsg'
Learn More

Tropaeolin Oo, Solstice, Flauka™

Product Code. 15613200 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15613200 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15613200 Supplier Honeywell-Fluka Supplier No. 310525G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Indicator, S. No.: 179

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 554-73-4
Packaging Glass bottle
Molecular Formula C18H14N3NaO3S
MDL Number MFCD00038130
UN Number NONH for all modes of transport
Beilstein 3831566
Quantity 5 g
InChI Key MLVYOYVMOZFHIU-UHFFFAOYSA-M
Physical Form Powder
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(NC2=CC=CC=C2)C=C1
IUPAC Name sodium;4-[(4-anilinophenyl)diazenyl]benzenesulfonate
Molecular Weight (g/mol) 375.38
PubChem CID 23668839
Formula Weight 375.38g/mol
Chemical Name or Material Tropaeolin Oo, Indicator, S. No.: 179
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.