missing translation for 'onlineSavingsMsg'
Learn More

Tropic Acid, TRC

Product Code. 17993495
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993495 10 g 10g
17921815 25 g 25g
17944365 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993495 Supplier Toronto Research Chemicals Supplier No. TRCT89262010G
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-63-6
Molecular Formula C9 H10 O3
Molecular Weight (g/mol) 166.17
Synonym (2RS)-3-Hydroxy-2-phenylpropanoic Acid, dl-Tropic Acid, Tropic Acid, Ipratropium Bromide Imp. C (EP), Hyoscyamine Sulphate Imp. A (EP), Hyoscine Butylbromide Imp. B (EP), Tropicamide Imp. C (EP), Hyoscine Hydrobromide Imp. D (EP), Atropine Imp. C (EP), (±)-α-(Hydroxymethyl)benzeneacetic acid, (±)-2-Phenyl-3-hydroxypropionic acid, (±)-3-Hydroxy-2-phenylpropionic acid, (±)-Tropic acid, 2-Phenyl-3-hydroxypropanoic acid, 2-Phenyl-3-hydroxypropionic acid, 2-Phenylhydracrylic acid, 3-Hydroxy-2-phenylpropionic acid, DL-Tropic acid, NSC 20990, α-(Hydroxymethyl)benzeneacetic acid
IUPAC Name 3-hydroxy-2-phenylpropanoic acid
SMILES OCC(C(=O)O)c1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H10O3/c10-6-8(9(11)12)7-4-2-1-3-5-7/h1-5,8,10H,6H2,(H,11,12)
Formula Weight 166.063
Percent Purity >95%
Chemical Name or Material rac Tropic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.