missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tropicamide, 99+%

Product Code. 15403779
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15403779 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15403779 Supplier Thermo Scientific Chemicals Supplier No. J61132.03
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1508-75-4 | C17H20N2O2

A muscarinic antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 g
Description White
Content And Storage 2°C to 8°C
Format Powder
For Use With (Application) A muscarinic antagonist
CAS 1508-75-4
Molecular Formula C17H20N2O2
Formula Weight 284.35g/mol
Synonym N-Ethyl-2-phenyl-N-(4-pyridylmethyl)hydracrylamide; Ro 1-7683
Melting Point 98°C
Product Type Signal Transduction Reagent
Form Powder
Assay ≥99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.