missing translation for 'onlineSavingsMsg'
Learn More

Tropinone, 99%

Product Code. 11371477
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11371477 100 g 100g
11351477 5 g 5g
11361477 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11371477 Supplier Thermo Scientific Alfa Aesar Supplier No. L02011.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 532-24-1 | C8H13NO | 139.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-24-1
Molecular Formula C8H13NO
Molecular Weight (g/mol) 139.20
MDL Number MFCD00005549
InChI Key QQXLDOJGLXJCSE-UHFFFAOYNA-N
Synonym tropinone, 8-methyl-8-azabicyclo 3.2.1 octan-3-one, 3-tropanone, tropanone, tropanon, tropinon, tropionone, 3-tropinone, 8-azabicyclo 3.2.1 octan-3-one, 8-methyl, 1.alpha.h,5.alpha.h-tropan-3-one
PubChem CID 79038
IUPAC Name 8-methyl-8-azabicyclo[3.2.1]octan-3-one
SMILES CN1C2CCC1CC(=O)C2

Specifications

Melting Point 40°C to 43°C
Boiling Point 227°C
Flash Point 90°C (194°F)
Odor Odorless
Quantity 100 g
Beilstein 2329
Formula Weight 139.2
Percent Purity 99%
Chemical Name or Material Tropinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.