missing translation for 'onlineSavingsMsg'
Learn More

Tropinone, 99%, Thermo Scientific Chemicals

Catalog Number 10055953
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
532-24-1
C8H13NO
139.20
MFCD00005549
QQXLDOJGLXJCSE-UHFFFAOYNA-N
tropinone, 8-methyl-8-azabicyclo 3.2.1 octan-3-one, 3-tropanone, tropanone, tropanon, tropinon, tropionone, 3-tropinone, 8-azabicyclo 3.2.1 octan-3-one, 8-methyl, 1.alpha.h,5.alpha.h-tropan-3-one
79038
8-methyl-8-azabicyclo[3.2.1]octan-3-one
CN1C2CCC1CC(=O)C2
This item is not returnable. View return policy
532-24-1
C8H13NO
139.20
MFCD00005549
QQXLDOJGLXJCSE-UHFFFAOYNA-N
tropinone, 8-methyl-8-azabicyclo 3.2.1 octan-3-one, 3-tropanone, tropanone, tropanon, tropinon, tropionone, 3-tropinone, 8-azabicyclo 3.2.1 octan-3-one, 8-methyl, 1.alpha.h,5.alpha.h-tropan-3-one
79038
8-methyl-8-azabicyclo[3.2.1]octan-3-one
CN1C2CCC1CC(=O)C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 41.0°C to 44.0°C
Color Yellow to Brown
Boiling Point 113.0°C (25.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 21,258
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol, ether, acetone, benzene, slightly soluble in chloroform
Formula Weight 139.2
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tropinone, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tropinone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'