missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Trovafloxacin mesylate
GREENER_CHOICE

Product Code. 18722694
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18722694

Brand: Tocris Bioscience™ 3863/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibiotic; inhibits bacterial DNA synthesis

Fluoroquinolone antibiotic. Inhibits bacterial DNA topoisomerase IV and DNA gyrase and forms a stable quinolone-DNA complex with these enzymes which reversibly inhibits DNA synthesis. Displays potent activity against gram-positive and gram-negative bacteria. Increases the production of mitochondrial NO in immortalized hepatocytes; also increases mitochondrial Ca2+. Inhibits Panx-1 (IC50 ∽ 4μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147059-75-4
Molecular Formula C21H19F3N4O6S

Specifications

Quantity 50 mg
Formula Weight 512.46
Purity >99%
Molecular Weight (g/mol) 512.46
Chemical Name or Material Trovafloxacin mesylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.