missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TRPC6-PAM-C20

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TRPC6-PAM-C20 is a selective TRPC6 positive allosteric modulator. Selectively activates TRPC6 over other TRP channels. Also activates TRPA1. Induces transient increase in intracellular Ca2+ in HEK cells expressing TRPC6 (EC50 = 2.37 μM). Potentiates OAG (1-oleoyl-1-acetyl-sn-glycerol)-induced Ca2+ influx in human platelets, and enhances OAG-induced platelet aggregation.
Specifications
Specifications
| Chemical Name or Material | 3-(3-,4-Dihydro-6,7-dimethoxy-3,3-dimethyl-1-isoquinolinyl)-2H-1-benzopyran-2-one |
| CAS | 667427-75-0 |
| Quantity | 50 mg |
| Molecular Formula | C22H21NO4 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction