missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ True North™ Mini-Cooler

Product Code. 16390676 Shop All Heathrow Scientific Products
Change view
Click to view available options
Array:
3 x 4
4 x 8
8 x 12
Color:
Blue
Green
Description:
Clear Lid
Gel Filled Lid
Withstand Temperature (Metric):
-20°C
0°C
Unit Size:
Each
Product Code. Array Color Description Withstand Temperature (Metric) unitSize
16390676 3 x 4 Green Clear Lid 0°C Each
16300686 4 x 8 Green Clear Lid 0°C Each
16310686 4 x 8 Green Gel Filled Lid 0°C Each
16320686 3 x 4 Blue Clear Lid -20°C Each
16330686 4 x 8 Blue Clear Lid -20°C Each
16340686 4 x 8 Blue Gel Filled Lid -20°C Each
16350686 8 x 12 Blue Clear Lid -20°C Each
7 options
This item is not returnable. View return policy

Product Code. 16390676

Brand: Heathrow Scientific™ HS120070

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The True North™ Mini Cooler protects temperature-sensitive samples at the bench or in the freezer. The Mini Cooler is ideal for keeping PCR™ reagents, enzymes, and other biochemical reagents cool while working at the bench.

  • Polycarbonate construction
  • Cooler has rubber non-slip feet
  • Gridded and keyed one-position lid for easy sample identification
  • Available with lid filled with insulating gel

Specifications

Color Green
Height (Metric) 10 cm
Height (English) 3.9 in.
Length (Metric) 14.5 cm
Length (English) 5.7 in.
Width (Metric) 10.5 cm
Width (English) 4.1 in.
Description Clear Lid
For Use With (Application) Tubes
Holds Tubes
Quantity 1 Ea.
Array 3 x 4
Material Polycarbonate
Type Cooler
Capacity (Metric) 0.5 to 2 mL
Withstand Temperature (Metric) 0°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.