missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Trypan Blue

Product Code. 10164110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10164110 25 g Glass bottle 25g
10174110 100 g Glass bottle 100g
15169213 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10164110 Supplier Thermo Scientific Chemicals Supplier No. 189350250
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 72-57-1 | C34H24N6Na4O14S4 | 960.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-57-1
Molecular Formula C34H24N6Na4O14S4
Molecular Weight (g/mol) 960.79
MDL Number MFCD00003969
InChI Key WMYVHJWZUUEZNE-ARWFNKCKSA-J
Synonym C.I. 23850, Direct Blue 14, Niagara Blue 3B
PubChem CID 9562061
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1

Specifications

Melting Point >280.0°C
Dye Content 60% min.
Infrared Spectrum Authentic
Packaging Glass bottle
UV Spectrometry Complies
Color Black/Brown to Blue
Solubility Information Solubility in water: 10g/L (25°C). Other solubilities: practically insoluble in alcohol, soluble in cellosolve, ethylene glycol, acids
Quantity 25 g
Physical Form Powder
IUPAC Name tetrasodium;(3Z)-5-amino-3-[[4-[4-[(2Z)-2-(8-amino-1-oxo-3,6-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate
Formula Weight 960.78
Chemical Name or Material Trypan Blue
Show More Show Less
compliance-icons
Product Identifier
  • Trypan Blue
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P233-Keep container tightly closed.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.