missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Trypan Blue, dye content >60%

Product Code. p-7024253
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11458517

Brand: Thermo Scientific Chemicals A18600.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72-57-1 | C34H24N6Na4O14S4 | 960.79 g/mol

Trypan blue is commonly used in microscopy (for cell counting) and in laboratory mice for assessment of tissue viability. Used in dye exclusion studies in cell culture. A viability dye for quantifying live versus dead cells with permeable membranes, blue. It is also used to identify islet cell cluster cell viability from caprine pancreatic samples. It is also useful to observe hyphea of fungi and Stramenopiles. Trypan blue solution may be used in trypan blue based cytotoxitiy and proliferation assays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trypan blue is commonly used in microscopy (for cell counting) and in laboratory mice for assessment of tissue viability. Used in dye exclusion studies in cell culture. A viability dye for quantifying live versus dead cells with permeable membranes, blue. It is also used to identify islet cell cluster cell viability from caprine pancreatic samples. It is also useful to observe hyphea of fungi and Stramenopiles. Trypan blue solution may be used in trypan blue based cytotoxitiy and proliferation assays.

Solubility
Soluble in water (10 mg/ml at 25°C), ethanol (0.6 mg/ml), and EGME (20 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-57-1
Molecular Formula C34H24N6Na4O14S4
Molecular Weight (g/mol) 960.79
MDL Number MFCD00003969
InChI Key WMYVHJWZUUEZNE-ARWFNKCKSA-J
PubChem CID 9562061
IUPAC Name tetrasodium;(3Z)-5-amino-3-[[4-[4-[(2Z)-2-(8-amino-1-oxo-3,6-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1

Specifications

Melting Point >300°C
Merck Index 14,9792
Solubility Information Soluble in water (10mg/ml at 25°C),ethanol (0.6mg/ml),and EGME (20mg/ml).
Formula Weight 960.82
Quantity 25 g
Chemical Name or Material Trypan Blue, dye content >60%
compliance-icons
Product Identifier
  • Trypan Blue
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Trypan Blue

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.