missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Trypsin (0.25%), phenol red
GREENER_CHOICE

Product Code. 11520626
Change view
Click to view available options
Quantity:
100 mL
20 x 100mL
Unit Size:
100mL
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11520626 100 mL 100mL
11530626 20 x 100mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11520626 Supplier Gibco™ Supplier No. 25050014
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Trypsin (0.25%), phenol red

Gibco™ Trypsin solutions are made from trypsin powder—an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength, it is widely used for cell dissociation, during routine cell culture passaging and primary tissue dissociation. We offer a variety of Gibco™ Trypsin and animal origin free TrypLE™ that feature:

  • Quality Testing
  • Documented Traceability
  • Dual-site cGMP Manufacturing



    Quality Testing
    Gibco™ Trypsin solutions are tested for pH, osmolality, sterility and performance. In addition, prior to manufacturing, the raw materials are tested for endotoxin, Mycoplasma, bacterial, fungal and viral contamination, as well as multiple activity assays, ash analysis and moisture analysis.

    Documented Traceability
    We can provide detailed documentation to meet your regulatory needs. Gibco™ Trypsin information available includes lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency.

    cGMP Manufacturing and Quality System
    Gibco™ Trypsin is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Trypsin solution made in our Grand Island, New York facility (15050-065). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ISO 13485 standards, FDA
    Content And Storage –5 to –20°C
    Description Trypsin, 100mL, 1 X Concentration, Liquid, 0.25% Dilution, 9002-07-7 CAS Number, With Phenol Red, No EDTA Chelators, Porcine Pancreas Source, Trypsin Reagent Solution, FDA and ISO 13485 Standards, Used for Mammalian Cell Culture, –5 to –20°
    Cell Type Mammalian Cells
    Concentration 1 X
    Form Liquid
    Product Type Cell Culture Dissociation Reagent
    With Additives Phenol Red
    Without Additives No EDTA
    For Use With (Application) Mammalian cell culture
    Quantity 100 mL
    Regulatory Status RUO
    Source Porcine pancreas
    Show More Show Less
    compliance-icons
    Product Identifier
    • Trypsin (1X), liquid
    Signal Word
    • Danger
    Hazard Category
    • Respiratory sensitiser Category 1
    Hazard Statement
    • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Precautionary Statement
    • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
    • P284-[In case of inadequate ventilation] wear respiratory protection.
    • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
    • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
    Supplemental information
    • MIXTURE LIST-Contain: trypsin

    For Research Use or Further Manufacturing. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.