missing translation for 'onlineSavingsMsg'
Learn More

Trypsin, 1-250 from Porcine Pancreas , MP Biomedicals, LLC

Product Code. 15782412 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
20 g
Packaging:
Glass
Unit Size:
20g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15782412 20 g Glass 20g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15782412 Supplier MP Biomedicals Supplier No. 0210313920

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trypsin, 1-250 from Porcine Pancreas has an activity >250 USP u/mg. Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues.

  • The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture
  • The 1:250 designation indicates the tryptic activity based on standardized testing
  • This ratio indicates that one part trypsin will digest 250 parts casein in the USP activity assay
  • Trypsin is composed of two subunits, a-trypsin and b-trypsin. a-Trypsin is composed of two peptide chains and b-trypsin is composed of one chain
  • Trypsin is used to re-suspend cells adherent to the cell culture dish wall during the process of harvesting cells
  • Trypsin can also be used to dissociate dissected cells
  • Trypsin can be used to break down casein in breast milk
  • Trypsin is commonly used in biological research during proteomics experiments to digest proteins into peptides for mass spectrometry analysis
  • Trypsin can also be used to dissolve blood clots in its microbial form and treat inflammation in its pancreatic form
  • One USP trypsin unit is the activity causing a change in absorbance of 0.003 per minute under the conditions specified
TRUSTED_SUSTAINABILITY

Specifications

CAS 7-7-9002
Color White
pH ∼8.0
Packaging Glass
Molecular Formula C35H47N7O10
Quantity 20 g
Synonym tryptase, trypsin, tryptase from human lung, tryptase from human lung, buffered aqueous solution, ≥5 units/mg protein, acetic acid; tert-butyl n-2s-1-2s-1-2s-5-diaminomethylideneamino-1-5-methyl-2-oxochromen-7-yl amino-1-oxopentan-2-yl amino-3-hydroxy-1-oxopropan-2-yl amino-1-oxo-3-phenylpropan-2-yl carbamate
InChI Key WGWZNYKOUXOZTC-UHFFFAOYNA-N
Molecular Weight (g/mol) 725.80
PubChem CID 5311489
compliance-icons compliance-icons
Product Identifier
  • Trypsin, 1-250
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: trypsin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.