missing translation for 'onlineSavingsMsg'
Learn More

Tryptamine, 98%

Product Code. 11996611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11996611 250 g Glass bottle 250g
10720891 10 g Glass bottle 10g
10427390 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11996611 Supplier Thermo Scientific Acros Supplier No. 186122500
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 61-54-1 | C10H12N2 | 160.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-54-1
Molecular Formula C10H12N2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00005661
InChI Key APJYDQYYACXCRM-UHFFFAOYSA-N
Synonym tryptamine, 2-1h-indol-3-yl ethanamine, 3-2-aminoethyl indole, 1h-indole-3-ethanamine, 2-3-indolyl ethylamine, indol-3-ethylamine, indole, 3-2-aminoethyl, 2-indol-3-yl ethylamine, 2-1h-indol-3-yl ethan-1-amine, tryptamin
PubChem CID 1150
ChEBI CHEBI:16765
IUPAC Name 2-(1H-indol-3-yl)ethanamine
SMILES NCCC1=CNC2=CC=CC=C12

Specifications

Melting Point 113.0°C to 117.0°C
Color White
Boiling Point 137.0°C (0.1 mmHg)
Flash Point 185°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 250 g
Beilstein 22,I,636
Merck Index 15,9976
Solubility Information Solubility in water: negligible. Other solubilities: soluble in ethanol and acetone, practically insoluble in ether, benzene and, chloroform
Formula Weight 160.22
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Tryptamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.