missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptic Digestion Kits

Product Code. 10688274 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Digestion and Guanidination Kit
Tryptic Digestion Kit
Quantity:
150 Reactions
90 Reactions
Unit Size:
1 set
90 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity unitSize
10688274 Tryptic Digestion Kit 150 Reactions 1 set
10371264 Digestion and Guanidination Kit 90 Reactions 90 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 10688274 Supplier Thermo Scientific™ Supplier No. 89871

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

Improve peptide digest quality and sensitivity of MS detection using these Thermo Scientific Digestion Kits for efficient and reproducible preparation of peptide digests.

Accurate identification and analysis of proteins is a central component of proteomic strategies for studying cellular functions and processes. Using mass spectrometry, proteins and relevant post-translational modifications can be accurately identified, but the method is highly dependent on the quality of the sample preparation. Thermo Scientific Pierce Digestion Kits provide enzymes and reagents for efficient, reproducible digestion and guanidination to improve peptide fragment ionization.

Sample preparation is one of the most variable and time-consuming steps in the analysis of proteins by MS, and the quality and reproducibility of sample extraction and preparation significantly impact the results. Occasionally, additional steps are required to improve sample quality, including digestion and recovery of peptide fragments from SDS-PAGE gels, or peptide guanidination to improve sequence coverage. Each kit has been optimized to produce high quality peptide digests for dependable proteomics analysis using mass spectrometry.

Pierce In-Solution Tryptic Digestion and Guanidination Kit

The Pierce In-Solution Tryptic Digestion and Guanidination Kit provides an optimized procedure and reagents for approximately 90 digests, each containing 0.025 to 10 μg of protein.

  • Complete digestion—trypsin completely digests protein ranging from 0.025 to 10 μg (∼90 reactions)
  • Specific—guanidination reaction produces little to no N-terminal modification
  • Convenient—kit includes reagents for digestion, reduction, alkylation, and guanidination
  • Efficient—protein can be digested and guanidinated in a single day

Trypsin specifically cleaves peptide bonds at the carboxyl side of arginine and lysine residues. The signals from the arginine-containing peptides are generally stronger, due to the more basic side chain. To enhance overall ionization, guanidination converts lysines to homoarginines and is specific for the amine of lysine, but may occur minimally at the amine of the peptide's N-terminus. This derivatization leads to an increase in the intensity of the lysine-containing peptides and improved sequence coverage overall.

Pierce In-Gel Tryptic Digestion Kit

The Pierce In-Gel Tryptic Digestion Kit provides a complete set of reagents for performing approximately 150 digestions on colloidal Coomassie- or fluorescent dye-stained protein bands. The kit includes modified porcine trypsin, de-staining buffers, reduction reagents, alkylation reagents, and digestion buffers, along with detailed and simple instructions.

  • Complete—includes all necessary reagents for gel destaining, reduction, alkylation, and tryptic digestion of proteins
  • Robust—optimized procedure and reagents produce reliable digestions and data generation using a wide range of conditions
  • Specific—contains highly purified and modified MS-grade trypsin with no chymotryptic activity and minimal autolytic activity

Two key sample preparation steps are the complete digestion of proteins into peptides and subsequent concentration and clean-up of peptides for mass spectrometry analysis. The Pierce In-Gel Tryptic Digestion Kit accomplishes the first of these steps for protein samples extracted from gel slices. Pierce C-18 Spin Columns and Tips accomplish the second step to clean-up the sample to be MS-ready.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Tryptic Digestion Kit
Content And Storage Upon receipt store Pierce Trypsin Protease, MS Grade at -20°C in a nonfrost-free freezer. Store all other components at 4°C. Product shipped on gel pack. See Important Product Information for additional storage and handling information for each component upon receipt.
Starting Material Gels, Protein Samples, Cell Lysate
Final Product Type Peptides
Detection Method Mass Spectrometry
Format Kit
Workflow Step Protein Digestion
Quantity 150 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.