missing translation for 'onlineSavingsMsg'
Learn More

Melford Trypto Soy Broth

Product Code. 30018867
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 30018867

Brand: Melford T48500500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dissolve 30 grams in 1 liter of distilled water.

Dissolve 30 grams in 1 liter of distilled water. Sterilize by autoclaving for 15 minutes.

Formula Per Liter:

Casein Digest Peptone: 17.0 g

Papaic Digest of Soybean Meal: 3.0 g

Dipotassium Phosphate: 2.5 g

Dextrose: 2.5 g

Sodium Chloride: 5.0 g

Specifications

Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.