missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Soya Agar EP/USP/JP/BP (Dehydrated)
GREENER_CHOICE

Product Code. 10483983
Change view
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
Unit Size:
3kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10483983 5 kg 5kg
10137562 500 g 500g
11973752 2.5 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10483983 Supplier Thermo Scientific™ Supplier No. CM0131T
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Grow a wide variety of microorganisms with this general purpose medium specified in international pharmacopoeia.

Grow a wide variety of microorganisms with Thermo Scientific ™ Oxoid ™ Tryptone Soya Agar (Dehydrated). A general purpose agar medium containing two peptones, Tryptone Soya Agar supports the growth of both aerobes and anaerobes, the latter being grown either in deep cultures or by incubation under anaerobic conditions. Tryptone Soya Agar is recommended as a reference medium when testing selective media to measure the degree of inhibition1.

Tryptone Soya Agar, also known as Casein Soya Bean Digest Agar, is a general purpose medium specified in pharmacopoeia and food testing methods for the growth of a wide variety of organisms. The product is tested to the pharmacopoeia2, 3, 4, 5 and is mainly used as reference medium when testing selective media.

  • Recommended as a reference medium for testing selective media
  • Conforms to formulations detailed in the International pharmacopoeia

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Tryptone Soya Agar
Quantity 5 kg

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.