missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Tryptophan hydroxylase 2 Antibody Blocking Peptide

Product Code. 18528502
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18528502 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18528502 Supplier Novus Biologicals™ Supplier No. NB10074555PEP
  • ONLINE EXCLUSIVE PRICE
    218.00 €
    205.80 € / 0.1mg
    Save 12.20 €
    5% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Gene ID (Entrez) 121278
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-74555
Gene Alias ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2
Gene Symbol TPH2
Quantity 0.1 mg
Research Category Lipid and Metabolism, Phospho Specific
Storage Buffer Peptide dissolved in dH20. Contains no BSA.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.