missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Tryptophan hydroxylase 2 Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 121278 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP1-77556. For further blocking peptide related protocol, click here . |
| Gene Alias | ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2 |
| Gene Symbol | TPH2 |
| Quantity | 0.1 mg |
| Research Category | Lipid and Metabolism, Phospho Specific |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?