Learn More
Heinz Herenz TS-Swab Alu-Amies
Product Code. 15962121
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 150
Product Code. Sterility Unit Size
15962121 Sterile Pack of 150
1 options
Product Code. 15962121 Supplier Heinz Herenz Supplier No. 1020020
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Sterile with Amies medium. Microbiological transport set. Aluminium applicator with cotton head, sterilized by radiation, and single packed in peel-pack
We offer a wide range of sterile and nonsterile swabs:
  • Wood, PS, PP, aluminium or paper applicator
  • Each in various lengths available
  • With cotton, viscose, polyester or foam head
  • Diameter of the heads: small, medium or large
  • This guarantees that the right instrument is available for every special application.
  • For example, the cotton-tipped swabs are recommended for use by ENT specialists.
  • Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
  • Only suitable for single use
  • Use by medical personnel only
Specifications
Product Type Liquid Amies Transport System
Format Swab
Media Type Liquid Amies
Sterility Sterile
Color White