Learn More
Thermo Scientific™ TSE Series Under Counter Stacking Kit
Product Code. 31740938 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Undercounter Refrigerator/Freezer
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31740938 Undercounter Refrigerator/Freezer Each
1 options
Product Code. 31740938 Supplier Thermo Scientific™ Supplier No. 507548SK
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stacking Kit Under Counter support bracket kit. Required for TSE Series Stackable Refrigerators and Freezers
TRUSTED_SUSTAINABILITY
Specifications
Type Support Bracket Kit
For Use With (Equipment) Undercounter Refrigerator/Freezer
Includes Front plate (1), Back plate (2), M5x15 Screws (3), T4x12 Screws (8)
Quantity 1 Kit