missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSP50 Rat anti-Human, Clone: 260606, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2430-SP
This item is not returnable.
View return policy
Description
TSP50 Monoclonal specifically detects TSP50 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| TSP50 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Cancer/testis antigen 20, CT20probable threonine protease PRSS50, EC 3.4.25.-, protease, serine, 50, Serine protease 50, testes-specific protease 50, Testis-specific protease-like protein 50, TSP50testes specific protease 50 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 29122 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunoprecipitation | |
| 260606 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PRSS50 | |
| Mouse myeloma cell line NS0-derived recombinant human TSP50 Ala40-Ala363 (predicted) | |
| 25 μg | |
| Primary | |
| Detects human TSP50 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactvity with recombinant mouse TSP50 is observed. | |
| Human | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?