Learn More
BD Vacutainer™ Luer Adapters
Product Code. 15120716
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 100
Product Code. Sterility Unit Size
15120716 Sterile Pack of 100
1 options
Product Code. 15120716 Supplier BD Supplier No. 367300
  • ONLINE EXCLUSIVE PRICE
    25.35 €
    24.93 € / Pack of 100
    Save 0.42 €
    1% Off
View all promotions
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ideal for use with any female luer fitting. BD Vacutainer™ Luer Adapters are sterile devices with a multi-sampling valve that are designed for use with a catheter or hypodermic needle to collect blood using BD Vacutainer™ blood collection tubes.
The BD Vacutainer™ system is a closed system optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process.
  • Provides sterile, secure and safe specimen sampling for fast, accurate and consistent results
  • Offers the security of a threaded, locking luer connection, replacing a luer-slip device
  • Latex-free and compatible with a female luer connection or needleless IV site designed for luer lock access
  • Used together with the BD Vacutainer™ single-use tube holder and evacuated blood collection tubes
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
For Use With (Application) Blood collection from a catheter, needleless IV site, hypodermic needle or any female luer fitting; For safe and easy subculturing
For Use With (Equipment) Vacutainer™ Single-use Tube Holders and Vacutainer™ Evacuated Blood Collection Tubes
Includes With luer adapter
Latex Free Yes
Product Type Tube Adapters and Holder
Sterility Sterile