missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Tube Closures

Product Code. 11758603
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
White
Yellow
Product Type:
Ribbed Closure Plug
Straight Plug
Winged Cap
For Use With (Equipment):
10 mm Tube
11 mm Tube
13 mm Tube
13.5 mm Tube
18.5 mm Tube
Quantity:
1000/Bag
1500 Bag
2000 Bag
2500 Bag
3000 Bag
4000 Bag
Unit Size:
Pack of 1000
Pack of 1500
Pack of 2000
Pack of 2500
Pack of 3000
Pack of 4000
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type For Use With (Equipment) Quantity Unit Size
11758603 Natural Ribbed Closure Plug 11 mm Tube 2500 Bag Pack of 2500
11592762 White Ribbed Closure Plug 11 mm Tube - Pack of 2500
11758593 White Ribbed Closure Plug 13.5 mm Tube 1500 Bag Pack of 1500
11748603 Yellow Ribbed Closure Plug 11 mm Tube 2500 Bag Pack of 2500
11778613 Red Straight Plug 18.5 mm Tube 1000/Bag Pack of 1000
11748593 White Ribbed Closure Plug 10 mm Tube - Pack of 3000
11798613 Green Ribbed Closure Plug 10 mm Tube 3000 Bag Pack of 3000
11768613 Red Straight Plug 13.5 mm Tube 2000 Bag Pack of 2000
11723313 Natural Winged Cap 13.5 mm Tube 1500 Bag Pack of 1500
11708623 Green Ribbed Closure Plug 11 mm Tube 2500 Bag Pack of 2500
11798603 Red Ribbed Closure Plug 10 mm Tube 3000 Bag Pack of 3000
11728603 Blue Ribbed Closure Plug 10 mm Tube 3000 Bag Pack of 3000
11708613 Red Ribbed Closure Plug 11 mm Tube 2500 Bag Pack of 2500
11718613 Red Ribbed Closure Plug 13.5 mm Tube 1500 Bag Pack of 1500
11738603 Yellow Ribbed Closure Plug 10 mm Tube 3000 Bag Pack of 3000
11778593 White Straight Plug 11 mm Tube 4000 Bag Pack of 4000
11502772 White Straight Plug 13.5 mm Tube - Pack of 2000
12113791 Blue Ribbed Closure Plug 13.5 mm Tube 1500 Bag Pack of 1500
11825853 Blue Ribbed Closure Plug 13 mm Tube - Pack of 2500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Product Code. 11758603 Supplier Gosselin™ Supplier No. BA11028
  • ONLINE EXCLUSIVE PRICE
    59.80 €
    45.78 € / Pack of 2500
    Save 14.02 €
    23% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suitable for different usage of tubes and applications. Gosselin™ Tube Closures are offered in straight, winged, skirted, or screw cap styles and are manufactured from PE plastic.

  • All closures vertically ribbed for easier grip
  • Protected with a polyethylene outer bag to prevent particle contamination
TRUSTED_SUSTAINABILITY

Specifications

Product Type Ribbed Closure Plug
Material LDPE
Color Natural
Quantity 2500 Bag
For Use With (Equipment) 11 mm Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.