Learn More
Syngene™ Small Wave UV Tube
Product Code. 12863998 Shop All Syngene Products
Change view
Click to view available options
Wavelength:
254nm
Unit Size:
Each
Product Code. Wavelength Unit Size
12863998 254nm Each
1 options
Product Code. 12863998 Supplier Syngene™ Supplier No. SWT8
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Syngene™ Small Wave UV Tube for use in UV Transilluminators.
  • For use with all GX and GE model transilluminators
  • 254nm wavelength
  • Small wave
Specifications
Intensity Fixed
Wavelength 254nm
Description Short wave UV tube
For Use With (Equipment) All GX/GE model transilluminators
Type UV Tube Short Wave