missing translation for 'onlineSavingsMsg'
Learn More

TPP™ TubeSpin™ Bioreactor Tube Racks

Product Code. 11341794
Change view
Click to view available options
Positions:
10 and 18
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions Unit Size
11341794 10 and 18 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11341794 Supplier TPP™ Supplier No. 99017
  • ONLINE EXCLUSIVE PRICE
    6.53 €
    5.34 € / Pack of 1
    Save 1.19 €
    18% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from polypropylene for strength and durability. TPP™ TubeSpin™ Bioreactor Tube Racks are designed for 15/50mL TPP™ TubeSpin™ Bioreactor tube.

Manufactured from polypropylene for strength and durability. TPP™ TubeSpin™ Bioreactor Tube Racks are designed for 15/50mL TPP™ TubeSpin™ Bioreactor tube.

  • Alphanumeric coding
  • Can be disassembled and stored to save space
  • Fits similarly designed test tubes (15 and 50mL)
  • Autoclavable

Specifications

Product Type Tube Rack
Autoclavable Autoclavable
Holds Tubes (15 and 50 mL)
Material Polypropylene
Positions 10 and 18
Temperature (Metric) Range -190°C to +121°C
Dimensions (L x W x H) 209 x 98 x62mm
Diameter (Metric) Opening 17.1 mm and 30 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.