Learn More
Palintest™ Tubetest COD Reagent Tube with Mercury For High Chloride Sample
Product Code. 11938937
Change view
Click to view available options
Range:
0 to 150mg/L
0 to 20000mg/L
Unit Size:
Pack of 24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
11938937 0 to 20000mg/L Pack of 24
11918937 0 to 150mg/L Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11938937 Supplier Palintest™ Supplier No. PL467
  • ONLINE EXCLUSIVE PRICE
    112.89 €
    77.00 € / Pack of 24
    Save 35.89 €
    31% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable for working with samples with very high chloride levels, this high chloride formulation (2M), is ideal for working in laboratory and other types of scientific environments.
  • The mercury free formulation is recommended for general use 
  • The mercuric sulfate formulation (M) is provided for use in standard procedures which require the suppression of interference due to moderate levels of chloride
  • The high chloride formulation (2M) provides much higher levels of chloride suppression and is suitable for working with samples with very high chloride levels
  • The new M/C formulation with mercuric sulfate is specifically for use in non-Palintest™ instruments
Specifications
For Use With (Application) High Chloride Samples Such as Chemical Oxygen Demand (COD)
For Use With (Equipment) Photometer or UV-Vis Spectrophotometer
Includes Chemical Oxygen Demand (COD)
No. of Tests 25
Range 0 to 20000mg/L
Type COD Reagent Tube