missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Tubetest COD Reagent Tube with Mercury For Moderate Chloride Sample

Product Code. 11956473
Change view
Click to view available options
Range:
0 to 150mg/L
0 to 20000mg/L
Unit Size:
Pack of 24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
11956473 0 to 20000mg/L Pack of 24
11938927 0 to 150mg/L Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11956473 Supplier Palintest™ Supplier No. PL466
  • ONLINE EXCLUSIVE PRICE
    93.77 €
    74.87 € / Pack of 24
    Save 18.90 €
    20% Off
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for use in standard procedures which require the suppression of interference due to moderate levels of chloride.

  • The mercury free formulation is recommended for general use
  • The mercuric sulfate formulation (M) is provided for use in standard procedures which require the suppression of interference due to moderate levels of chloride
  • The high chloride formulation (2M) provides much higher levels of chloride suppression and is suitable for working with samples with very high chloride levels
  • The new M/C formulation with mercuric sulfate is specifically for use in non-Palintest™ instruments

Specifications

For Use With (Application) Moderate Chloride Samples Such as Chemical Oxygen Demand (COD)
For Use With (Equipment) Photometer or UV-Vis Spectrophotometer
Includes Chemical Oxygen Demand (COD)
No. of Tests 25
Range 0 to 20000mg/L
Type COD Reagent Tube
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • COD 20000/M Solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.