missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Tubetest COD Reagent Tube with Mercury

Product Code. 11908937
Click to view available options
Range:
0 to 2000mg/L
0 to 400mg/L
Unit Size:
0.72 L
Pack of 24
This item is not returnable. View return policy

Product Code. 11908937

Brand: Palintest™ PL462

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Practical and convenient method for testing Chemical Oxygen Demand (COD) using the Palintest™ COD Tubetests system.

  • The Palintest™ COD Tubetests system offers a method which is both practical and convenient
  • The reagents are supplied premixed and with a minimum of potentially hazardous chemicals
  • The test is simply carried out by adding the sample to the tube, closing the cap and then digesting for two hours at 150°C
  • The tubes are then cooled and the test colour measured using a Palintest™ photometer

Specifications

For Use With (Application) Moderate Chloride Samples Such as Chemical Oxygen Demand (COD)
For Use With (Equipment) Photometer or UV-Vis Spectrophotometer
Includes Chemical Oxygen Demand (COD)
No. of Tests 25
Range 0 to 400mg/L
Type COD Reagent Tube
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P314-Get medical advice/attention if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P363-Wash contaminated clothing before reuse.
  • P391-Collect spillage.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.