Learn More
Palintest™ Tubetest Mercury Free COD Reagent Tube
This mercury-free COD reagent tube is ideal for measuring low chloride samples such as Chemical Oxygen Demand (COD).
Brand: Palintest™ PL450
Code : 04
Additional Details : CAS Number : 7664-93-9 Weight : 0.48600kg Transport : UN number : 3264 Chem class : 8 Pack group : II
Description
- Mercury-free COD reagent tube for measuring low chloride samples such as Chemical Oxygen Demand (COD)
- For use with Photometer or UV-Vis Spectrophotometer equipment
- Type: COD Reagent Tube
- Range: 0 to 150mg/L
Specifications
Low Chloride Samples Such as Chemical Oxygen Demand (COD) | |
Chemical Oxygen Demand (COD) | |
25 | |
COD Reagent Tube |
Photometer or UV-Vis Spectrophotometer | |
25 | |
0 to 150mg/L |
Safety and Handling
- COD 150 Solution
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P363-Wash contaminated clothing before reuse.
- P391-Collect spillage.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, silver sulphate, potassium dichromate