missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBGCP2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 468.00 €
Specifications
| Antigen | TUBGCP2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TUBGCP2 Polyclonal antibody specifically detects TUBGCP2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TUBGCP2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 10844 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Gamma-ring complex protein 103 kDa, gamma-tubulin complex component 2, gamma-tubulin complex protein 2, GCP-2, GCP2hGrip103, h103p, hGCP2, hSpc97, MGC138162, SPBC97, Spc97p, Spindle pole body protein Spc97 homolog, tubulin, gamma complex associated protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human TUBGCP2 (NP_006650.1). MSEFRIHHDVNELLSLLRVHGGDGAEVYIDLLQKNRTPYVTTTVSAHSAKVKIAEFSRTPEDFLKKYDELKSKNTRNLDPLVYLLSKLTEDKETLQYLQQNAKERAELAAAAVGSSTTSINVPAAASKISMQELEELRKQLGSVATGSTLQQSLELKRKMLRDKQNKKNSGQHLPIFPAW | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title