missing translation for 'onlineSavingsMsg'
Learn More

BOLA Tubing Connectors Elbow

Product Code. 10439381
Click to view available options
Diameter (Metric) Outer:
4.5 mm
6.8 mm
9 mm
11 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10439381

Brand: BOLA D57408

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Tubing Connectors Elbow PTFE

  • Elbow-shaped fitting made of PTFE with two connectors for elastic tubing (e.g. Viton™, Tygon™, silicone).
  • Universal chemical resistance, the product is only exposed to PTFE.

Specifications

Certifications/Compliance FDA
Autoclavable Autoclavable
Diameter (Metric) Outer 11 mm
Overall Length (Metric) 28.5 mm
Color White
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.