missing translation for 'onlineSavingsMsg'
Learn More

BOLA Tubing Connectors T

Product Code. 10359371
Click to view available options
Diameter (Metric) Outer:
6.8 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10359371

Brand: BOLA D57704

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Tubing Connectors T PTFE

  • T-shaped fitting made of PTFE with three connectors for elastic tubing (e.g. Viton™, Tygon™, silicone).
  • Universal chemical resistance
  • The product is only exposed to PTFE.

Specifications

Certifications/Compliance FDA
Autoclavable Autoclavable
Diameter (Metric) Outer 6.8 mm
Overall Length (Metric) 22.5 mm
Color White
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.