Learn More
Vici Valco Instruments™ Tubing Cutter
Product Code. 10338739
Change view
Click to view available options
Type:
Tubing Cutter
Unit Size:
Each
Product Code. Type Unit Size
10338739 Tubing Cutter Each
1 options
Product Code. 10338739 Supplier Vici Valco Instruments™ Supplier No. JR796
  • ONLINE EXCLUSIVE PRICE
    108.00 €
    18.28 € / Pack of 1
    Save 89.72 €
    83% Off
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The pliers type of cutter can be used for quick cuts for non-critical applications or at hard to get places.
  • For easy cutting of 1/16 in. tubing off a coil
  • Reach “hard-to-get” places in an HPLC system
  • Delivered in a plastic sheath
Specifications
Type Tubing Cutter
Description Easy cutting of 1/16 in. tubing off a coil