missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Tapered Hose Coupling

Polyethylene disconnectors provide a quick, flexible, and safe alternative to expensive adapters. Kartell™ Tapered Hose Couplings tightly join glass, rubber, or plastic hoses together.

Brand:  Kartell™ 7053600

Product Code. 12549756

  • ONLINE EXCLUSIVE PRICE
    19.00 €
    18.42 € / Pack of 10
    Save 0.58 €
    3% Off
In Stock
Edge
Add to basket

Description

Description

  • Useful for joining hoses of glass, rubber or plastic
  • Connector fits tightly together
  • Replaces expensive adapters
Specifications

Specifications

CE/IVD
Bore Size 7.3 mm
12/13/14 mm
10
Natural
12 to 14 mm
Polyethylene
Glass, rubber, or plastic hoses
7.3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.