Learn More
Hamilton™ Tubing Retention Clips
Product Code. 12790388
Click to view available options
Quantity:
5/Pk.
Unit Size:
Pack of 5
Product Code. 12790388
Brand: Hamilton™ 88990
Holds tubing in place with beakers and other vessels.
  • 5 clips per pack
  • 12 or 18 gauge
Specifications
For Use With (Application) Holds tubing
Product Type Retention Clips
Quantity 5/Pk.
For Use With (Equipment) Microlab 500 A, B, and C Series