missing translation for 'onlineSavingsMsg'
Learn More

(+)-Tubocurarine Chloride Hydrochloride Pentahydrate, TRC

Product Code. 30454256
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454256 25 mg 25mg
30456875 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30454256 Supplier Toronto Research Chemicals Supplier No. TRCT94762025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (+)-Tubocurarine chloride hydrochloride pentahydrate
CAS 6989-98-6
Molecular Formula C37 H41 N2 O6 . Cl . Cl H . 5 H2 O
Quantity 25 mg
Synonym 13H-4,6:21,24-Dietheno-8,12-metheno-1H-pyrido[3',2':14,15][1,11]dioxacycloeicosino[2,3,4-ij]isoquinolinium','2,3,13a,14,15,16,25,25a-octahydro-9,19-dihydroxy-18,29-dimethoxy-1,14,14-trimethyl-','chloride','hydrochloride','pentahydrate','(13aR,25aS)-, Tubocuraranium','7',12'-dihydroxy-6,6'-dimethoxy-2,2',2'-trimethyl-','chloride','hydrochloride','pentahydrate, (+)-Tubocurarine chloride hydrochloride pentahydrate, Tubarine pentahydrate, d-Tubocurarine dichloride pentahydrate
InChI Formula InChI=1S/C37H40N2O6.2ClH.5H2O/c1-38-14-12-24-19-32(42-4)33-21-27(24)28(38)16-22-6-9-26(10-7-22)44-37-35-25(20-34(43-5)36(37)41)13-15-39(2,3)29(35)17-23-8-11-30(40)31(18-23)45-33;;;;;;;/h6-11,18-21,28-29H,12-17H2,1-5H3,(H-,40,41);2*1H;5*1H2/t28-,29+;;;;;;;/m0......./s1
SMILES O.O.O.O.O.Cl.[Cl-].COc1cc2CC[N+](C)(C)[C@@H]3Cc4ccc(O)c(Oc5cc6[C@H](Cc7ccc(Oc(c1O)c23)cc7)N(C)CCc6cc5OC)c4
IUPAC Name (13aR,25aS)-2,3,13a,14,15,16,25,25a-Octahydro-9,19-dihydroxy-18,29-dimethoxy-1,14,14-trimethyl-13H-4,6:21,24-dietheno-8,12-metheno-1H-pyrido[3',2':14,15][1,11]dioxacycloeicosino[2,3,4-ij]isoquinolinium chloride hydrochloride pentahydrate
Molecular Weight (g/mol) 771.72
Formula Weight 770.2948
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.