Learn More
SCHOTT AG Lighting and Imaging™ Tubular diffuser Only for VisiLED Ring Lights
Shop All Schott Products
Click to view available options
Type:
Tubular Diffuser
Unit Size:
Pack of 20
Description
VisiLED diffuser made of improved materials is designed specifically for use only with VisiLED brightfield ring lights (S80-65N-2).
- Prevents LED reflections
- High transmission and perfect haze of more than 99%
- Includes adapting thread (M90 x 1)
- Controller needs to be ordered separately
Specifications
Specifications
| Type | Tubular Diffuser |
| For Use With (Application) | A Wide Variety of Tasks From Life Science to Industrial Applications |
| For Use With (Equipment) | EasyLED Ring light (600 200), EasyLED Ringlight Plus (600 300), VisiLED range S80-25 and S80-55 Ringlights (400225 and 400255) manufactured from August 2023 |