missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TUG 891

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent FFA4 (GPR120) agonist (pEC50 values are 7.36 and 7.77 for human and mouse GPR120 respectively). Selective for GPR120 over free fatty acid receptors (pEC50 = 4.19 for FFA1; displays no activity at FFA2 or FFA3). Cell permeable.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 364.41 |
| Purity | >99% |
| Molecular Weight (g/mol) | 364.416 |
| Chemical Name or Material | TUG 891 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction