missing translation for 'onlineSavingsMsg'
Learn More

Tungsten sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)

Product Code. 15424865
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424865 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15424865 Supplier Thermo Scientific Alfa Aesar Supplier No. 041640.KS
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-33-7 | W | 183.84 g/mol

Tungsten was used extensively for the filaments of old-style incandescent light bulbs, its alloys are used in many high-temperature applications, such as arc-welding electrodes and heating elements in high-temperature furnaces. Some bacteria use tungsten in an enzyme to reduce carboxylic acids to aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
CAS 7440-33-7
Molecular Formula W
MDL Number MFCD00011461
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
Solubility Information Insoluble in water.
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES [W]
IUPAC Name tungsten
Molecular Weight (g/mol) 183.84
PubChem CID 23964
ChEBI CHEBI:27998
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Tungsten sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.