missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten(V) dichloride triethoxide, 10% w/v in ethanol, Thermo Scientific™
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Spécification
| Color | Blue |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Linear Formula | WCl2(OC2H5)3 |
| UN Number | UN2924 |
| Formula Weight | 389.94 |
| Concentration or Composition (by Analyte or Components) | 10% w/v in ethanol |
| Odor | Alcohol-like |
| Sensitivity | Moisture sensitive |
| Chemical Name or Material | Tungsten(V) dichloride triethoxide |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu