missing translation for 'onlineSavingsMsg'
Learn More

Tungsten(VI) Oxide, TRC

Product Code. 30450277
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450277 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450277 Supplier Toronto Research Chemicals Supplier No. TRCT8972605G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tungsten trioxide
CAS 1314-35-8
Quantity 5 g
Molecular Formula WO3
Synonym Tungsten oxide (WO3), Avantama P10, C.I. 77901, CQ 8116, CQ 81G16, CQM 30, Ditungsten hexaoxide, FI-WO 3, Renecat, SII 30, Tritungsten nonaoxide, Tungsten oxide, Tungsten oxide (W2O6), Tungsten oxide (W3O9), Tungsten oxide (W4O12), Tungsten trioxide, Tungsten trioxide (WO3), Tungsten(VI) oxide, Tungstic anhydride, Tungstic oxide, Tungstic oxide (WO3), WWO 03PB, YMDS 06
InChI Formula InChI=1S/3O.W
SMILES O=[W](=O)=O
IUPAC Name trioxotungsten
Molecular Weight (g/mol) 231.84
Formula Weight 231.9357
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.