missing translation for 'onlineSavingsMsg'
Learn More

Tungsten wire, chromium plated, 1.8mm (0.07in) min dia, W 96.5+%, Cr 99.888% (metals basis)

Product Code. 11319457
Click to view available options
Quantity:
10 cm
5 x 10 cm
10 x 10 cm
Unit Size:
10cm
Each
This item is not returnable. View return policy

Product Code. 11319457

Brand: Thermo Scientific Alfa Aesar 041040.BS

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Evaporation source, contacts and coatings

Evaporation source, contacts and coatings

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tungsten wire, chromium plated
Name Note ≤1.8mm (0.07 in.) dia., thick: 1.3mm (0.05 in.), Cr layer: ≤0.01 in. thick
Quantity 10 cm
Form Wire
Assay Percent Notes W ≥96.5%, Cr 99.888% (metals basis)
MDL Number MFCD00011461

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.