missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Turbidity Standard/Kit

Product Code. 10350605
Change view
Click to view available options
For Use With (Equipment):
Hach Turbidimeters
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10350605 Hach Turbidimeters Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10350605 Supplier Cole-Parmer™ Supplier No. 2659405
  • ONLINE EXCLUSIVE PRICE
    1115.94 €
    722.29 € / Pack of 1
    Save 393.65 €
    35% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with: HACH Turbidity meters

TRUSTED_SUSTAINABILITY

Specifications

Type Turbidity Standard Kit
For Use With (Equipment) Hach Turbidimeters
compliance-icons
Product Identifier
  • StablCal sealed vials calibration kits
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: methenamine; hexamethylenetetramine , methenamine; hexamethylenetetramine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.