missing translation for 'onlineSavingsMsg'
Learn More

TPP Techno Plastic Products AG Turbo-Fix Consumable Stabifix

Product Code. 17297016
Change view
Click to view available options
Turbo-fix, pipette aid, yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17297016 Turbo-fix, pipette aid, yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17297016 Supplier TPP Techno Plastic Products AG Supplier No. 94750
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Turbo-Fix consumable Stabifix nose piece

  • Turbo-Fix
  • Pipette aid
  • Ideal for controled sensitive pipetting speed
  • Lightweight but still strong

Specifications

Compatible Tips Serological pipettes mouthpiece
Color Gray
For Use With Serological Pipettes, Nose Piece for Turbo Fix Pipette Aid
Model Nose piece
Number of Channels 1
Sterility Sterile
Volume (Metric) Size of serological pipette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.